Z743914
ART® SoftFit L Reload System
HLT 1000L, size 1000 μL, reload insert, non-sterile
Synonym(s):
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip
About This Item
reload insert
sterility
non-sterile
feature
HLT 1000L
reload insert
packaging
pkg of 4X768 ea
manufacturer/tradename
Molecular BioProducts 3782-RI
technique(s)
PCR: suitable
size
1000 μL
volume
1000 μL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Reload towers present an eco-friendly packaging solution.
- Reduces plastic waste and optimizes storage space requirements.
- Compatible with SoftFit-L empty racks.
- Color-coded inserts enable quick and effortless identification of tip volume, streamlining your workflow.
- Reload towers are an environmentally-friendly packaging option that reduces plastic waste and storage space.
- Compatible with SoftFit-L empty racks.
- Color-coded inserts allows quick identification of tip volume.
- Free of DNase, RNase, DNA, ATP, and endotoxin.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service