Key Documents
Z743272
Julabo® adaptors
barbed fittings, for 10 mm i.d. tubing, pack of 2
Synonym(s):
barbed adaptor
Select a Size
2 EA
$69.50
$69.50
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
2 EA
$69.50
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
$69.50
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
packaging
pack of 2
manufacturer/tradename
Julabo 8970447
General description
For use with Julabo® circulators, 2 barbed fittings for tubing 10 mm ID. For use with M16x1 fittings. Requires a female M16x1 cap nut.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)