Key Documents
Z743209
Julabo® bath accessories
hollow polyproplylene balls, diam. 20 mm, for use in Julabo CORIO and Pura water baths, pack of 1000 pieces
Select a Size
1 EA
$296.00
$296.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$296.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
$296.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
packaging
pack of 1000 pieces
manufacturer/tradename
Julabo 8970010
General description
Place the hollow polyproplylene balls in a thermostat bath to form an even covering layer, exclusively for use in waterr. The floating balls reduce: heat loss, evaporation loss (up to 90%), oxygen input, odors, light exposure and are food-safe up to 100°C.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)