Key Documents
Z743203
Julabo® bath carrier trays for SW22/SW23
universal basic tray for assembling spring clamps
Select a Size
1 EA
$762.00
$762.00
Estimated to ship onApril 06, 2025
Select a Size
Change View
1 EA
$762.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
$762.00
Estimated to ship onApril 06, 2025
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970630
General description
Basic stainless steel tray for assembling Erlenmeyer flasks from 25 to 1000 mL, including spring clamps. Arrange springs to hold flasks in preferred arrangement. For use in shaking water bath models SW22, SW23.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)