Key Documents
Z743201
Julabo® bath carrier trays for SW22/SW23
basic tray for assembling spring clamps
Select a Size
1 EA
$470.00
$470.00
Estimated to ship onApril 10, 2025
Select a Size
Change View
1 EA
$470.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
$470.00
Estimated to ship onApril 10, 2025
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970620
General description
Basic stainless steel tray for assembling spring clamps. For use with Julabo® shaking water bath models SW22, SW23. Mix and match combinations of spring clamps Z743249 (8970601) through Z557471 (8970609) install on the basic tray.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)