Key Documents
Z743134
Julabo® cryo-compact circulator
CF40, capacity 5.5 L, temperature -40-150 °C, precision: ±0.03, AC/DC input 115 V AC, US 3-pin plug (5-15p)
About This Item
UNSPSC Code:
41103706
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9400340
temperature measuring range
-40-150 °C, precision: ±0.03
capacity
5.5 L
General description
The new CF series offers compact refrigerated/heating circulators. With small overall dimensions the units can even be placed on a very small surface or within a complex installation. All models offer a heating capacity of 2 kW and safety class 3 according to DIN12876-1. The maximum ambient temperature of +40°C as well as a ventilation air cooling allow installing the units very closely side by side with other units or directly under a fume hood. Furthermore all the models feature pump connections for external temperature applications and a bath opening for temperature control of small objects directly in the circulator bath. Wetted parts are made of high quality stainless steel or plastic. The instruments provide a reliable Microprocessor electronics with high temperature stability as well as warning and safety functions.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)