Z742986
Julabo® CORIO™ C open heating bath circulator
C-B17, capacity 13 L (17 L), stainless steel bath (Insulated), temperature 20-100 °C, AC/DC input 230 V AC, Schuko plug
All Photos(1)
About This Item
Recommended Products
material
stainless steel bath (Insulated)
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9011417
temperature measuring range
20-100 °C
capacity
13 L (17 L)
external W × L × H
38 cm × 40 cm × 47 cm
opening W × L × H
30 cm × 18 cm × 20 cm
General description
Open heating bath circulators are designed for internal applications in the circulator bath.
CORIO C open heating bath circulators in this range are equipped with bath tanks made of Polycarbonat or stainless steel.
Available accessories include a variety of test tube racks, immersion-height adjustable platforms and cooling coils. The circulator can be removed for easy cleaning of the bath tanks.
CORIO C open heating bath circulators in this range are equipped with bath tanks made of Polycarbonat or stainless steel.
Available accessories include a variety of test tube racks, immersion-height adjustable platforms and cooling coils. The circulator can be removed for easy cleaning of the bath tanks.
Legal Information
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.