Key Documents
Z742527
Block for digital dry bath and MyBlock
Holds 1 x 96-well PCR plate
Select a Size
1 EA
$407.00
$407.00
Estimated to ship onMay 28, 2025
Select a Size
Change View
1 EA
$407.00
About This Item
UNSPSC Code:
41102404
NACRES:
NB.22
$407.00
Estimated to ship onMay 28, 2025
feature
Holds 1 x 96-well PCR plate
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific BSWPCR2
technique(s)
PCR: suitable
General description
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Legal Information
MyBlock is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)