Key Documents
Z742522
Block for digital dry bath and MyBlock
Holds 20 x 12-13 mm tubes
Select a Size
1 EA
$156.00
$156.00
Estimated to ship onMay 26, 2025
Select a Size
Change View
1 EA
$156.00
About This Item
UNSPSC Code:
41102404
NACRES:
NB.77
$156.00
Estimated to ship onMay 26, 2025
feature
Holds 20 x 12-13 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific BSW13
General description
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Legal Information
MyBlock is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)