Key Documents
Z742519
Block for digital dry bath and MyBlock
Holds 12 x 5.0 mL tubes
Select a Size
1 EA
$156.00
$156.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$156.00
About This Item
UNSPSC Code:
41102404
NACRES:
NB.77
$156.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
Holds 12 x 5.0 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific BSW5MT
General description
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Legal Information
MyBlock is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)