Key Documents
Z742314
2MAG MIX 1 XL single point stirrers up to 40 L
AC/DC input 100-240 V, US 3-pin plug
Synonym(s):
high volume stirrer, induction plate, induction stirrer, laboratory stirrer, magnetic stir plate, magnetic stirrer
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
Pricing and availability is not currently available.
material
stainless steel housing
stainless steel plate
feature
positions 1
0 ‑ 240 S Variable Intervals (2 step)
Power Setting 5/10/15/20 W ((4-step)
4 step)
packaging
pack of 1 ea
manufacturer/tradename
2mag 30201
parameter
-10-50 °C temp. limit (at 95 percent humidity)
70-2000 rpm speed
W × D × H
225 mm × 280 mm × 35 mm
plate size × W × L
× 220 mm × 250 mm
stirrer volume
1-40 L , per stirring position
weight
~4.0 kg
General description
Ultra flat, strong magnetic stirrer with large set-up area and fully automatic stirring bar monitoring mixWatch, up to 40 litres stirring volume, also suitable for highly viscous media, very strong magnetic coupling, therefore predestined for magnetic power transmission over wide distances or for thick vessel bottoms.
ntegrated controller with strong 20 watts stirring power, 4-step power setting for high power with regard to large and viscous stirring volumes and reduced power for the warming-free continuous operation, SoftStart-function (adjustable, 4 steps) for reliable catching/centering and safe accelerating of the stirring bar at different volumes and viscosities, adjustable interval times for automated restart of the stirrer after max. 240 seconds.
Choose from one of the most recent versions:
Certificates of Analysis (COA)