Skip to Content
Merck
All Photos(1)

Key Documents

Z742308

Rocking platform for RotoBot Mini

size mini, size 6 in. × 5.5 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$211.00

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$211.00

About This Item

UNSPSC Code:
41103811
NACRES:
NB.61

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

size

mini
6 in. × 5.5 in.

feature

includes flat mat

manufacturer/tradename

Benchmark Scientific R4040-RP

Looking for similar products? Visit Product Comparison Guide

General description

For rocking applications, this platform with a non-slip rubber mat is an accessory for the Rotobot and Rotobot Mini rotating mixers. Once attached, simply select the "PP" mode to turn the rotator into a platform rocker.

Other Notes

For use with the Rotobot Mini Rotating Mixer Z742297/8/9 (Benchmark Scientific number R4040).

Legal Information

Rotobot is a trademark of Benchmark Scientific, Inc.

accessory


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service