Key Documents
Z742262
DURAN® TILT Light Shield
white silicone, includes GL 56 bottle tags
Synonym(s):
292435601
Select a Size
4 EA
$99.30
$99.30
Estimated to ship onMarch 28, 2025
Select a Size
Change View
4 EA
$99.30
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
$99.30
Estimated to ship onMarch 28, 2025
material
white silicone
feature
includes GL 56 bottle tags
packaging
pack of 4
manufacturer/tradename
DURAN® 292435601
General description
The DURAN TILT light shield is a white silicone sleeve that covers 94% of the bottle surface. The sleeve has a number of protective functions. It blocks damaging UV light, protects the surface from damage, and facilitates safer handling. Silicone provides a much better grip than glass, especially when the bottle has been sprayed with 70% alcohol for surface disinfection.
Features and Benefits
• Manufactured from silicone• The white DURAN® TILT Light Shield is a silicone sleeve that covers 94 % of the bottle surface and blocks damaging ultraviolet light• Protects the glass surface from damage• Facilitates safer handling• Silicone provides a much better grip than glass especially when the bottle has been sprayed with 70 % alcohol for surface disinfection
Other Notes
Includes four GL 56 Bottle Tags - orange, yellow, blue, and purple
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)