Key Documents
Z741120
Sigma-Aldrich
Aldrich® Essentials round media storage bottle
wide mouth, capacity 2000 mL, GL 45 polypropylene cap with pour ring
Select a Size
4 EA
$262.00
$262.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
4 EA
$262.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
$262.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass bottle
polypropylene cap
round bottle
Joint
Joints threaded neck (GL45)
sterility
non-sterile
feature
graduations
with cap
packaging
pkg of 4 ea
manufacturer/tradename
Sigma-Aldrich
capacity
2000 mL
General description
Borosilicate glass, wide mouth, media/reagent bottles with polypropylene GL 45 thread screw cap with pour ring. Large marking spot and reference graduations.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)