Key Documents
Z740680
Sigma-Aldrich
Aldrich® Essentials filter flask, with side arm
capacity 250 mL
Select a Size
12 EA
$308.00
$308.00
Estimated to ship onMay 29, 2025
Select a Size
Change View
12 EA
$308.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$308.00
Estimated to ship onMay 29, 2025
material
borosilicate glass flask
flat bottom flask
Joint
joint stopper6
packaging
pkg of 12 ea
capacity
250 mL
General description
Clear borosilicate glass Erlenmeyer style flask with side arm for connecting to vacuum or aspirator line. Used for filtration with Buchner funnels, Gooch crucibles, etc. Beaded rim for rubber gromets or filter adapters or rubber stoppers.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Which type of flask is suitable for a #6 stopper?
    1 answer
    1. The flask Z740680 is not sized for a #6 stopper, as its opening is too wide, causing the stopper to fall through. However, the item CLS70340250 does fit a #6 stopper.
      Helpful?