Skip to Content
Merck
All Photos(1)

Key Documents

Z687766

Replacement NMR needle with cone tip for eVol® electronic syringe

for 5 μL, L × O.D. 11.5 cm × 0.5 mm, gauge size 25

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
$62.80

$62.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
2 EA
$62.80

About This Item

UNSPSC Code:
41121705
NACRES:
SB.32

$62.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

gauge size

25

manufacturer/tradename

SGE 036922

L × O.D.

11.5 cm × 0.5 mm

Looking for similar products? Visit Product Comparison Guide

General description

The eVol® NMR electronic syringe facilitates in-tube sample mixing and enables recovery of precious samples allowing them to be stored in alternative vessels (such as inexpensive vials) rather than being left in valuable tubes. The cone tip replacement needles come in multiple length and gauge options to fit your specific needs.

Legal Information

eVol is a registered trademark of SGE Analytical Science Pty Ltd.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service