Key Documents
Z685461
Vacuubrand chemistry pumping units, automatic control, Model PC 610 NT
AC/DC input 230 V AC, European 2-pin plug
Synonym(s):
evacuation pump, evaporation pump, evaporator pump
About This Item
UNSPSC Code:
41105101
NACRES:
NB.79
manufacturer/tradename
Vacuubrand 737100
parameter
1.5 mbar vacuum
3.4-3.8 m3/h pumping speed
General description
Chemistry pumping units of this series are well proven for vacuum generation and control for many evaporation processes. The popular two-stage MZ 2C NT chemistry diaphragm pump is the heart of these pumping units, frequently used for medium sized vacuum applications involving "common" solvents.
The pumping units are equipped with a CVC 3000 vacuum controller with a solenoid valve for electronic vacuum control. The exhaust waste vapor condenser is outstandingly efficient and compact. It provided near 100 percent recovery of solvents, for economical recycling and environmental protection.
  • Optimized vacuum even with gas ballast
  • Easily operated controller with clear text menus with integrated venting valve
  • Chemistry design flow control valve with large cross section for unrestricted performance
  • Whisper quiet and very low vibration
  • Excellent environmental friendliness due to efficient solvent recovery
Choose from one of the most recent versions:
Certificates of Analysis (COA)