Z683574
Syringe PP/PE without needle
luer slip tip, centered, capacity 5 mL, graduated, 0.2 mL, non-sterile
Synonym(s):
polypropylene syringes
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
sterility
non-sterile
feature
graduated 0.2 mL
packaging
pkg of 100 ea bulk packed
capacity
5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
These polypropylene syringes with Luer tips, may be used with any size needle. The rigid polyethylene plungers of these disposable syringes are slightly oversized, creating a tight seal against the walls of the semi-flexible PP barrel, as well as providing an exceptionally smooth draw. No contaminating lubricants added.
Syringes are individually packaged (sterile) except for bulk packed products; these are supplied in plastic bags (non-sterile).
Order needles separately.
Syringes are individually packaged (sterile) except for bulk packed products; these are supplied in plastic bags (non-sterile).
Order needles separately.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service