Key Documents
Z682691
Ace drying flask, pear-shaped, with side indents
capacity 1000 mL, joint: ST/NS 29/32, glass
Synonym(s):
Aldrich® drying flask, pear-shaped, with side indents
Select a Size
1 EA
$392.00
$392.00
Estimated to ship onMay 24, 2025
Select a Size
Change View
1 EA
$392.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$392.00
Estimated to ship onMay 24, 2025
material
glass
pear-shaped flask
single-neck
Joint
joint: ST/NS29/32
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Büchi equivalent, 00453
capacity
1000 mL
General description
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)