Key Documents
Z682640
Ace evaporating flask, pear-shaped
capacity 2000 mL, joint: ST/NS 29/42, poly-coated, heavy wall
Synonym(s):
Aldrich® evaporating flask, pear-shaped, bottom flask, flask round, round flask
Select a Size
1 EA
$246.00
$246.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$246.00
About This Item
UNSPSC Code:
41121804
NACRES:
VB.02
$246.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
pear-shaped flask
single-neck
Joint
joint: ST/NS29/42
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Ace Glass 3990-124
capacity
2000 mL
General description
These pear-shaped evaporating flasks can be used as boiling type flasks for rotary evaporators.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)