Key Documents
Z682497
Ace vapor duct tube, for condenser "V" assembly
joint: ST/NS 24/40, L 45 mm, fits Büchi® models R-114/R-144
Select a Size
1 EA
$287.00
$287.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$287.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$287.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
manufacturer/tradename
Büchi equivalent, 32335
L
45 mm
Related Categories
General description
The vapor duct tube accessories are for Büchi condenser "V" assemblies, and will fit either Büchi Rotavapor model R-210/R-215 or R-114/R-144.
Combi-clips: not included.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)