Key Documents
Z682195
Ace receiving flask adapter
joint: ST/NS 24/40, fits Büchi® models R-114/R-144
Select a Size
1 EA
$463.00
$463.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$463.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
$463.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
manufacturer/tradename
Büchi equivalent, 23747
Related Categories
General description
The receiving flask adapter fits condenser assemblies "C", "CR", "A", and "V" for Büchi® evaporator models R-200/205. Adapter clip: not included.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)