Key Documents
Z679933
Bio-Pure pipetting reservoir
capacity 25 mL, with divider
Synonym(s):
multichannel pipette reservoir, reagent reservoir
Select a Size
100 EA
$202.00
$202.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
100 EA
$202.00
About This Item
UNSPSC Code:
41121522
NACRES:
NB.01
$202.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
PET (Bio-degradable)
V-bottom bottom
white
sterility
sterile
packaging
pack of 100 ea
manufacturer/tradename
Diversified Biotech REBP-2000
capacity
25 mL
General description
Bio-Pure Reagent Reservoirs are made from recaputured PET that does not rely on additional fossil fuel-derived petrochemicals. In addition, reservoirs are sealed in bio-degradable bags and packed in recycled cardboard cases. As a result, Bio-Pure Reservoirs are a more eco-friendly alternative. "Trough within a trough" maximizes reagent recovery. A wide base and reinforced sidewalls avoids spills and provides stability. Gamma irradiated RNASE/DNASE free
Features and Benefits
  • Constructed from biodegradable PET material for environmentally conscious disposal.
  • Specialized "trough within a trough" design for optimal reagent recovery.
  • Pour-off spouts at all four corners and a wide base for improved stability and spill prevention.
  • Sterilized for safe and contamination-free usage.
Legal Information
Bio-Pure is a trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)