Z678104
Finnpipette® F3 variable volume pipette
volume 2-20 μL
Select a Size
$652.00
Estimated to ship onMay 30, 2025
Select a Size
About This Item
$652.00
Estimated to ship onMay 30, 2025
Recommended Products
feature
color-coded turquoise
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Finn 4640020
volume
2-20 μL
Related Categories
General description
- Double action pipetting button with very light and smooth pipetting action
- Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
- Big display with white numbers on black background for increased visibility
- Color coded for easy tip volume recognition
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)