Key Documents
Z677922
Finntip® Flex pipette tips
Finntip Flex 1000, sterile
Select a Size
960 EA
$199.00
$199.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
960 EA
$199.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
sterility:
sterile
feature:
barrier: no
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Finn 94060713
$199.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
blue
colorless tip
sterility
sterile
feature
barrier: no
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Finn 94060713
maximum volume
1 mL
volume range
100-1000 μL
General description
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Other Notes
Browse all pipette tips
Legal Information
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)