Z677698
Vitoject™ fluorocarbon rubber gloves
size S (8)
Synonym(s):
chemically resistant gloves, hand protection against aggressive chemicals
About This Item
Recommended Products
material
black (fluorocarbon rubber)
black gloves
size
S (8)
sterility
non-sterile
feature
CE compliant
latex free
powder-free: no
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
packaging
pack of 1 pair
manufacturer/tradename
KCL 890
L × thickness
350 mm × 0.7 mm
Looking for similar products? Visit Product Comparison Guide
General description
Manufactured with the ecologically beneficial and modern Vulcoject-process technology (injection molding).
- Very good protection against extremely aggressive chemicals
- 100% gastight
- Excellent wearing comfort
- Solvent free by eco-friendly injection molding process
- CE compliant
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service