Skip to Content
Merck
All Photos(2)

Key Documents

Z676675

ART® barrier tip

maximum volume 100 μL, Tip design ART 100, Hinged rack, low retention: no

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 960 EA
$1,040.00

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 X 960 EA
$1,040.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
100 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
low retention: no
Hinged rack
Tip design ART 100
packaging:
pack of 4800 ea (50 hinged racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2065-HR

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier
low retention: no
Hinged rack
Tip design ART 100

packaging

pack of 4800 ea (50 hinged racks of 96 tips)

manufacturer/tradename

Molecular BioProducts 2065-HR

technique(s)

PCR: suitable

maximum volume

100 μL

volume

100 μL

Looking for similar products? Visit Product Comparison Guide

General description

The self-sealing ART barrier tips are supplied in environmentally friendly packaging. The sterile, barrier tip reload system combines individually packaged blister tubs made of recycled plastic and new hinged racks. The push down blister tub easily inserts the ART barrier tips into the hinged racks providing stackable, space saving racks that offer easy, one-handed use. Empty hinged rack part numbers allow for easy conversion to the ART Reload System.

Manufactured according to the industry′s highest quality standards, ART tips are certified as RNase, DNase and Pyrogen free, and are ideal for the pipetting of radioactive samples, nucleic amplification procedures, or any applications where critical sample handling is required.

Available as reload inserts consisting of sterile pre-loaded blister tubs of 96 tips with or without hinged racks. The empty hinged racks are also available separately.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service