Key Documents
Z648914
Sigma-Aldrich
Suba-Seal® septa
blue rubber, Suba-Seal® 37, fits neck I.D., 19 mm
Select a Size
100 EA
$253.00
$253.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
100 EA
$253.00
About This Item
UNSPSC Code:
41121800
eCl@ss:
32119211
NACRES:
NB.35
$253.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
blue rubber
feature
fits neck I.D. 19 mm
manufacturer/tradename
Suba-Seal® 37
General description
Well-known as the best quality rubber turn-over closure with serrated stopper, Suba-Seal brand is available exclusively from Sigma-Aldrich. Ideal for air and moisture-sensitive chemistry procedures. After piercing with non-coring needle, rubber closes puncture, providing air and moisture tight seal to protect contents of vessel or container from atmosphere.
This closure has turnover flange molded to grip outside of container neck promoting a double seal in conjunction with annular serrations on hollow plug. Annular serrations depress themselves against the inside wall of the bottle neck, making each serration a suctions sealing point.
Storage Note for Natural Rubber Septa: Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.
Note: For extra security when using large bore transfer needles, fasten septum to bottle or flask opening with a wrap-it tie. See ties and accessory tool listings.
Legal Information
Suba-Seal is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)