Key Documents
Z648183
Grid coating pen
provides a thin layer of adhesive
Select a Size
1 EA
$156.00
$156.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$156.00
About This Item
UNSPSC Code:
41100000
NACRES:
MA.01
$156.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
manufacturer/tradename
1556
General description
A light touch with this pen will provide a thin layer of adhesive on the grid. The layer will dry in 1-2 minutes at room temperature and is then ready to receive the sections. Pen may also be used as pre-treatment before coating grids with formvar or other thin films.
Choose from one of the most recent versions:
Certificates of Analysis (COA)