Key Documents
Z645761
Chemical spill bucket
Synonym(s):
chemical cleanup, spill cleanup, spill safety
Select a Size
1 KIT
$345.00
$345.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 KIT
$345.00
About This Item
UNSPSC Code:
47131905
NACRES:
NB.35
$345.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
manufacturer/tradename
Clear-spill CK4
availability
not available in USA and Canada, ROW
General description
Kit designed for small to medium spills, ideal for laboratories. Contains 20 pads, 3 x 8 cm by 1.2 m socks, 2 x 30 cm by 35 cm cushion, bags & ties with instructions supplied in a durable container with a snap close lid. Kit will absorb 31 litres.
PLEASE NOTE: KIT NO LONGER CONTAINS PPE
Choose from one of the most recent versions:
Certificates of Analysis (COA)