Key Documents
Z645184
IKA® A 11.1 spare beater
A 11.1 spare beater, 1/cs
Select a Size
1 EA
$133.00
$133.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$133.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
$133.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
stainless steel
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 2904600
suitability
suitable for (pulverizing substances with a Mohs hardness up to 6)
General description
Stainless steel replacement bat for use with the IKA A11 Basic Analytical Mill, for breaking substances up to Mohs hardness 6.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)