Z610291
Hirschmann® volumetric pipette, Class AS
capacity 20 mL , 0.03 mL accuracy
Select a Size
$103.00
Select a Size
About This Item
$103.00
Recommended Products
material
soda-lime glass
feature
class AS
packaging
pack of 6 ea
manufacturer/tradename
Hirschmann 1340165
availability
not available in USA and Canada
H
510 mm
capacity
20 mL , 0.03 mL accuracy
color
amber graduations
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service