Key Documents
Z568708
Sigma-Aldrich
Aldrich® combustion apparatus platinum sample carrier
pkg of 1 ea
Synonym(s):
Combustion Apparatus
Select a Size
1 EA
$2,510.00
$2,510.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$2,510.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$2,510.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 1 ea
General description
Has a basket on lower end that holds the paper-wrapped sample.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)