Z567191
Ace round-bottom pressure flask
with Ace-Thred 15 PTFE front-seal plug, capacity 250 mL
Synonym(s):
Round-Bottom Flask
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
round bottom flask
single-neck
sterility
non-sterile
feature
closure type PTFE plug seal
packaging
pack of 1
manufacturer/tradename
Ace Glass 841511
capacity
250 mL
diam.
82 mm
General description
Heavy-wall borosilicate glass flasks are pressure rated to 60 psig at 120°C. The rounded bottom facilitates use in heating mantles and for other round-bottom flask applications. Flasks are supplied complete with Ace-Thred PTFE front seal bushing and FETFE® O-ring.
Replacement FETFE O-rings:
Ace-Thred #Product #
7 Z504661
15 Z504858
25 Z505056
14 Z504742
210 Z505021
220 Z505110
CAUTION: This glassware is not guaranteed against breakage caused by pressure or vacuum. These flasks should never be used if scratched or otherwise damaged.
Replacement FETFE O-rings:
Ace-Thred #Product #
7 Z504661
15 Z504858
25 Z505056
14 Z504742
210 Z505021
220 Z505110
CAUTION: This glassware is not guaranteed against breakage caused by pressure or vacuum. These flasks should never be used if scratched or otherwise damaged.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.