Z563315
Plas-Labs basic multi-station glove box
AC/DC input 240 V AC, Euro plug
Synonym(s):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Plas-Labs 818-GBB/EXP
H × W × D
28 in. × 60 in. × 38 in., inside dimensions
31 in. × 76 in. × 31 in., outside dimensions
weight
250 lb
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Price includes shipping
The Plas-Labs basic multi-station glove boxes are engineered to fit general isolation applications in chemical, pharmaceutical, biotech and waste management laboratories. They can be easily modified for specific uses in isolating sensitive research studies from a hostile exterior environment. The basic multi-station glove box is portable, lightweight and self-contained.
- Clear, one piece acrylic top with molded white base
- Double layered, closed cellular neoprene gaskets for airtight seal between top and base
- Adjustable SS clamp compensate for wear
- Two pairs of glove ports for two or more operators working in tandem.
- 8 in. O.D. ports with ambidextrous Hypalon® gloves with SS O-rings
- Transparent transfer chamber (12 L × 11 in i.d.) with adjustable vacuum gauge
- Three tier suspended (sliding) shelves are spaced 9 in. apart, shelves are made of 9.5 mm clear acrylic.
- White levelling tray for transferring liquids through the transfer chamber.
- Four purge valves, two on transfer chamber, two on main chamber
- Multiple electrical outlet strip, CE UL and CSA approved
Legal Information
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service