Key Documents
Z561894
Sigma-Aldrich
Aldrich® glass beaker with handle
capacity 300 mL, borosilicate glass
Synonym(s):
beaker mug
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
borosilicate glass
feature
graduations
manufacturer/tradename
Sigma-Aldrich
capacity
300 mL
General description
Made from 100% borosilicate glass with white, easy to read, graduations on back. With a 300 mL capacity, the large side handle stays cool when hot liquids are inside. These beaker mugs are heatproof, ovenproof, microwave and dishwasher safe.
The attached handle eliminates the need for clumsy gripping devices when handling hot liquids. Beakers are made of lightweight glass that is durable and has excellent heat transfer properties for rapid heating and cooling.
Other Notes
Browse all glass beakers
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)