Key Documents
Z560812
Sigma-Aldrich
Maxisafe safety utility knife
includes safety spring
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
General description
A safety spring provides for automatic blade "shoot-back" into the handle even in response to any uncontrolled movement.
Legal Information
Maxisafe is a trademark of Martor USA
Choose from one of the most recent versions:
Certificates of Analysis (COA)