Key Documents
Z557609
Sigma-Aldrich
Aldrich® pear-shaped splash-guard adapter
capacity 250 mL, female joint: ST/NS 24/40, male joint: ST/NS 24/40
Select a Size
1 EA
$251.00
$251.00
Estimated to ship onApril 16, 2025
Select a Size
Change View
1 EA
$251.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$251.00
Estimated to ship onApril 16, 2025
Joint
female joint: ST/NS24/40
male joint: ST/NS24/40
description
anti-climb
capacity
250 mL
General description
Application: for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping. Tapered sidewalls eliminate hold up common to round style adapters and permits continuous washing with condensed solvent.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)