Key Documents
Z553336
Sigma-Aldrich
Washer for NMR tubes with J. Young valve
top joint: ST/NS 19/26, bottom joint: ST/NS 24/40
About This Item
UNSPSC Code:
47131800
NACRES:
SB.32
Joint
bottom joint: ST/NS24/40
top joint: ST/NS19/26
General description
The inverted 5 mm NMR tube is sealed into the washer with a ground-glass stopper. Applied vacuum sprays solvent inside and against the closed end of tube for thorough cleaning.
Fits flask with standard taper 24/40 joint.
Choose from one of the most recent versions:
Certificates of Analysis (COA)