Key Documents
Z551600
Chemat compact hotplate
AC/DC input 220 V AC, European 2-pin plug
About This Item
UNSPSC Code:
41102406
NACRES:
NB.79
Pricing and availability is not currently available.
sterility
non-sterile
feature
CE compliant
manufacturer/tradename
Chemat KW-4AH-220
Chemat
availability
not available in EU
temperature measuring range
120-660 °F, resolution: 1 °C
parameter
50-350 °C temp. range
W × L
6 in. × 6 in.
General description
The portable design is convenient to be used in conjunction with the spin-coater. Has digital display and internal thermocouple.
Choose from one of the most recent versions:
Certificates of Analysis (COA)