Z551228
Graduated Valumetric™ bottles
black phenolic cap (with rubber liner), capacity 60 mL (2 oz)
Synonym(s):
W216979
All Photos(1)
About This Item
Recommended Products
material
black phenolic cap (with rubber liner)
clear bottle
round bottle
soda-lime glass bottle (Type III)
wide-mouth bottle
Joint
Joints threaded neck (38-400)
feature
with cap
packaging
pkg of 48 ea
manufacturer/tradename
WHEATON® W216979
capacity
60 mL (2 oz)
diam. × H
42 mm × 83 mm
General description
Ideal for use as sample bottles. They are also useful for dry powders, soil samples, and specimen storage. All bottles are made of clear, Type III soda lime glass.
Features and Benefits
• Clear, USP Type III soda-lime glass• Graduated in mL and ounces• Easy removal of contents• Available with caps attached or bulk packs without caps• HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification• Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)
Legal Information
Valumetric is a trademark of Wheaton Industries, Inc.
WHEATON is a registered trademark of Wheaton Industries, Inc.