Key Documents
Z550310
Sigma-Aldrich
Aldrich® Hirsch funnel
joint: ST/NS 14/20, porosity 1.0-1.6 μm
Select a Size
1 EA
$194.00
$194.00
Estimated to ship onApril 17, 2025
Select a Size
Change View
1 EA
$194.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
$194.00
Estimated to ship onApril 17, 2025
Joint
joint: ST/NS14/20
pore size
1.0-1.6 μm porosity
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)