Key Documents
Z549827
Sigma-Aldrich
Aldrich® jacketed three-neck flask
half jacket, capacity 2,000 mL, center joint: ST/NS 29/42
Synonym(s):
boiling flask, flask round, flask round bottom
Select a Size
1 EA
$2,930.00
$2,930.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$2,930.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$2,930.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
round bottom
three-neck
three-neck flask
Joint
center joint: ST/NS29/42
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
2,000 mL
General description
Unique design which incorporates a flush-closing PTFE drain valve eliminating unwanted dead space.
Features:
  • Eliminates dead space for complete stirring of flask contents
  • Allows for complete draining without breaking down flask set-up
  • Liquid cooled or heated with liquid or steam
  • 24/40 side joints
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)