Key Documents
Z531634
Sigma-Aldrich
Tubing Adapters with Torion Connection
O.D. 3/16 in., Straight, male joint: ST/NS 19/22
Select a Size
1 EA
$128.00
$128.00
Estimated to ship onApril 13, 2025
Select a Size
Change View
1 EA
$128.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$128.00
Estimated to ship onApril 13, 2025
Joint
male joint: ST/NS19/22
O.D.
3/16 in.
4.8 mm
Related Categories
General description
The unique Torion connection ensures an airtight seal between tubing and reaction vessels when transferring air-sensitive materials.
5/16 in. O.D fits a #8 Torion, 1/2 in. O.D. fits a #14 Torion, and 3/16 in. O.D fits a #5 Torion.
  • Hard-wall transfer tubing (e.g. PP or PTFE) is inserted through the threaded polypropylene bushing and PTFE-coated sealing ring of the adapter
  • Turn the bushing to make an airtight compression seal with the tubing
  • Use the optional solid-glass plug to make an airtight Torion seal when the transfer tubing is not in use
  • O.D. refers to the tubing outer diameter
Legal Information
Torion is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)