Key Documents
Z531227
Sigma-Aldrich
Aldrich® coiled condenser
jacketed, L 100 mm, joint: ST/NS 14/20
Select a Size
1 EA
$466.00
$466.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$466.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$466.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS14/20
L
100 mm
Related Categories
General description
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)