Key Documents
Z530387
Sigma-Aldrich
Aldrich® short-path distillation head
Joints: ST/NS 14/20 (2), thermometer joint: ST/NS 10/18
Select a Size
1 EA
$132.00
$132.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
$132.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
$132.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Joint
thermometer joint: ST/NS10/18
Joints: ST/NS14/20 (2)
immersion depth
50 mm
Related Categories
General description
Vigreux indents prevent bumping of fine or lightweight column packing into the condenser or fraction cutter when column is flooded.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)