Z529567
Silver Shield® gloves
size M
Synonym(s):
Antimicrobial Gloves, Protective Gloves, Silver-Infused Gloves
About This Item
Recommended Products
material
PE and EVOH
size
M
sterility
non-sterile
feature
disposable
latex free
powder-free
packaging
pkg of 10 pair
manufacturer/tradename
North SSG-9
L × thickness
16 in. (40.6 cm) × 2.7 mil
color
silver
Looking for similar products? Visit Product Comparison Guide
General description
Gloves feature a five-layer fabric compacted to a thickness of 2.7 mil. Laminating a high-energy polymer (EVOH) with a low-energy polymer (PE) creates a fabric impervious to at least 90% of all organic chemicals for four hours at 35°C. Each layer acts as a barrier against various epoxies and industrial chemicals.
- Lightweight; reduces hand fatigue without sacrificing dexterity
- Economical - Can be used as a disposable glove, without the need for cleaning and re-cycling
- Does not contain latex or chemical accelerators - Little chance of allergic reactions
- Ambidextrous, hypoallergenic
- Can be used as a secondary inner glove - Where the dangers of mechanical damage to gloves are high
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service