Key Documents
Z510351
Sigma-Aldrich
Vigreux distilling column
bottom joint: ST/NS 14/20
Select a Size
1 EA
$205.00
$205.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$205.00
About This Item
UNSPSC Code:
40161600
NACRES:
NB.43
$205.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS bottom14/20
Related Categories
General description
Vigreux indents run the entire length of the column and act as the packing for the column.
Choose from one of the most recent versions:
Certificates of Analysis (COA)