Key Documents
Z507857
Round-bottom flask with side port
capacity 5 mL
Select a Size
1 EA
$75.70
$75.70
Estimated to ship onMay 29, 2025
Select a Size
Change View
1 EA
$75.70
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$75.70
Estimated to ship onMay 29, 2025
material
borosilicate glass
round bottom flask
single-neck
sterility
non-sterile
feature
closure type Screw cap
packaging
pack of 1
manufacturer/tradename
Ace Glass 9592-35
capacity
5 mL
General description
Single neck flask has threaded <ST/>14/10 joint, and an additional side port with PTFE-faced silicone rubber septum for easy retrieval or injection access. Supplied with holed caps and FETFE® O-rings.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)