Key Documents
Z502413
Sigma-Aldrich
Glindemann PTFE sealing rings
(for 29/26, 29/42, 29/32 joints)
Synonym(s):
ptfe ring, ptfe sealing
Select a Size
1 PKG
$170.00
$170.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 PKG
$170.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
$170.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint 29/26
joint 29/32
joint 29/42
packaging
pkg of 50 ea
pkg of
manufacturer/tradename
Glindemann K29
temperature measuring range
≤250 °C
General description
  • This narrow ring seal is an alternative to a PTFE joint sleeve
  • Greaseless protection for air- and moisture- sensitive compounds
  • Can be used for prolonged storage under vacuum
  • Slide easily onto medium and full length taper joints
  • Can be used with Keck® clip and similar metal joint clips
Legal Information
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)